Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-propanediol-d6 | 284474-77-7 | MFCD01074057 | 99.3% | 82.13 g/mol | C3H2D6O2 | 10 MG
SDP

Catalog No. 5000300903
Encompass_Preferred

1,3-Propanediol-d6 is the deuterium-labeled isotopologue of 1,3-propanediol supplied as a high-purity research standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry, NMR reference, and isotope-tracing studies.

  • Isotopically labeled with six deuterium atoms (d6) for mass-shifted analysis.
  • High purity suitable for analytical applications (reported 99.29%).
  • Molecular weight 82.13 g/mol, molecular formula C3H2D6O2.
  • Available in small pack sizes for trace-level and standard preparation.
  • Suitable as an internal standard for MS, NMR, and tracer studies.

Catalog No. 50-003-00903 Supplier Medchemexpress LLC Supplier No. HYW017758S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Propanediol-d6 is the deuterium-labeled isotopologue of 1,3-propanediol supplied as a high-purity research standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry, NMR reference, and isotope-tracing studies.

  • Isotopically labeled with six deuterium atoms (d6) for mass-shifted analysis.
  • High purity suitable for analytical applications (reported 99.29%).
  • Molecular weight 82.13 g/mol, molecular formula C3H2D6O2.
  • Available in small pack sizes for trace-level and standard preparation.
  • Suitable as an internal standard for MS, NMR, and tracer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.