Learn More
Medchemexpress LLC 1,3-propanediol-d6 | 284474-77-7 | MFCD01074057 | 99.3% | 82.13 g/mol | C3H2D6O2 | 100 MG
SDP
Catalog No. 5000301634
Encompass_Preferred
1,3-Propanediol-d6 is the deuterium-labeled analogue of 1,3-propanediol (CAS 284474-77-7), supplied as a colorless to light yellow liquid. It is used as a stable-isotope standard and reagent for analytical and synthetic workflows, including mass spectrometry, NMR, and isotope-tracing studies. Reported molecular formula C3H2D6O2 and molecular weight 82.13 g/mol.
  • Deuterium-labeled analogue for isotope-dilution and tracer studies.
  • High purity (reported 99.29%) suitable for analytical applications.
  • Liquid form, colorless to light yellow, convenient for solution preparation.
  • Compatible with mass spectrometry and NMR workflows.
  • Available in milligram-scale quantities for laboratory use.
Catalog No. 50-003-01634 Supplier Medchemexpress LLC Supplier No. HYW017758S100MG
1,3-Propanediol-d6 is the deuterium-labeled analogue of 1,3-propanediol (CAS 284474-77-7), supplied as a colorless to light yellow liquid. It is used as a stable-isotope standard and reagent for analytical and synthetic workflows, including mass spectrometry, NMR, and isotope-tracing studies. Reported molecular formula C3H2D6O2 and molecular weight 82.13 g/mol.
  • Deuterium-labeled analogue for isotope-dilution and tracer studies.
  • High purity (reported 99.29%) suitable for analytical applications.
  • Liquid form, colorless to light yellow, convenient for solution preparation.
  • Compatible with mass spectrometry and NMR workflows.
  • Available in milligram-scale quantities for laboratory use.