Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-propanediol-d6 | 284474-77-7 | MFCD01074057 | 99.3% | 82.13 g/mol | C3H2D6O2 | 5 MG
SDP

Catalog No. 5000301763
Encompass_Preferred

1,3-Propanediol-d6 is a deuterium-labeled analogue of 1,3-propanediol supplied as an isotope-labeled reagent for research applications. It is used as an internal standard or tracer in analytical techniques such as mass spectrometry and isotope tracing. The compound has formula C3H2D6O2, a molecular weight of 82.13 g/mol, and CAS 284474-77-7; follow recommended storage conditions to preserve isotopic integrity.

  • Deuterium-labeled internal standard for analytical assays.
  • Molecular formula C3H2D6O2 and molecular weight 82.13 g/mol.
  • High isotopic and chemical purity (99.3%).
  • Suitable for mass spectrometry and tracer experiments.
  • Intended for research use only; not for human consumption.
  • Recommended cold storage to maintain stability and isotopic composition.

Catalog No. 50-003-01763 Supplier Medchemexpress LLC Supplier No. HYW017758S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,3-Propanediol-d6 is a deuterium-labeled analogue of 1,3-propanediol supplied as an isotope-labeled reagent for research applications. It is used as an internal standard or tracer in analytical techniques such as mass spectrometry and isotope tracing. The compound has formula C3H2D6O2, a molecular weight of 82.13 g/mol, and CAS 284474-77-7; follow recommended storage conditions to preserve isotopic integrity.

  • Deuterium-labeled internal standard for analytical assays.
  • Molecular formula C3H2D6O2 and molecular weight 82.13 g/mol.
  • High isotopic and chemical purity (99.3%).
  • Suitable for mass spectrometry and tracer experiments.
  • Intended for research use only; not for human consumption.
  • Recommended cold storage to maintain stability and isotopic composition.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.