Learn More
Medchemexpress LLC 1,3-propanediol-d6 | 284474-77-7 | MFCD01074057 | 99.3% | 82.13 g/mol | C3H2D6O2 | 50 MG
SDP
Catalog No. 5000299718
Encompass_Preferred
1,3-Propanediol-d6 is the deuterium-labeled analogue of 1,3-propanediol used as a stable isotope reference standard and analytical reagent. Supplied as a liquid with documented molecular weight and density, it is intended for research applications requiring isotopically labeled standards.
  • High isotopic enrichment and >99% chemical purity.
  • Suitable for mass spectrometry and NMR applications.
  • Useful as an internal or external standard in quantitative analysis.
  • Available in small research-grade package sizes for analytical use.
  • Stable liquid form with documented physical properties for handling.
Catalog No. 50-002-99718 Supplier Medchemexpress LLC Supplier No. HYW017758S50MG
1,3-Propanediol-d6 is the deuterium-labeled analogue of 1,3-propanediol used as a stable isotope reference standard and analytical reagent. Supplied as a liquid with documented molecular weight and density, it is intended for research applications requiring isotopically labeled standards.
  • High isotopic enrichment and >99% chemical purity.
  • Suitable for mass spectrometry and NMR applications.
  • Useful as an internal or external standard in quantitative analysis.
  • Available in small research-grade package sizes for analytical use.
  • Stable liquid form with documented physical properties for handling.