Learn More
Medchemexpress LLC 1,3-propanediol-d8 | 285978-25-8 | MFCD01074016 | 98.9% | 84.14 | C3D8O2 | 5 MG
SDP
Catalog No. 5000300904
Encompass_Preferred
1,3-Propanediol-d8 is the deuterium-labeled isotopologue of 1,3-propanediol used as an internal standard and analytical reference for mass spectrometry, isotope tracing, and other research applications. It has molecular formula C3D8O2, molecular weight 84.14, and CAS 285978-25-8.
  • Deuterium-labeled standard for quantitative analysis by GC-MS and LC-MS.
  • High isotopic enrichment for accurate isotope tracing experiments.
  • High chemical purity suitable for analytical workflows.
  • Available in small quantities that are convenient for method development.
  • Compatible with common sample preparation and derivatization protocols.
Catalog No. 50-003-00904 Supplier Medchemexpress LLC Supplier No. HYW017758S15MG
1,3-Propanediol-d8 is the deuterium-labeled isotopologue of 1,3-propanediol used as an internal standard and analytical reference for mass spectrometry, isotope tracing, and other research applications. It has molecular formula C3D8O2, molecular weight 84.14, and CAS 285978-25-8.
  • Deuterium-labeled standard for quantitative analysis by GC-MS and LC-MS.
  • High isotopic enrichment for accurate isotope tracing experiments.
  • High chemical purity suitable for analytical workflows.
  • Available in small quantities that are convenient for method development.
  • Compatible with common sample preparation and derivatization protocols.