Learn More
Medchemexpress LLC 1,3-propanediol-d8 | 285978-25-8 | MFCD01074016 | 98.9% | 84.14 g·mol⁻¹ | C3D8O2 | 1 MG
SDP
Catalog No. 5000301661
Encompass_Preferred
1,3-Propanediol-d8 is the deuterium-labeled analogue of 1,3-propanediol used as an isotope-labeled standard and reagent in analytical and synthetic research. It offers high isotopic enrichment and is provided in small, well-characterized quantities suitable for calibration, tracer studies, and method development.
  • Deuterium-labeled analogue for isotope tracing and standards.
  • High purity suitable for analytical applications (listed as 98.9%).
  • Suitable for GC-MS, NMR, and mass spectrometry workflows.
  • Provided in milligram-scale packages for standards and calibration.
  • Comes with documentation such as data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-003-01661 Supplier Medchemexpress LLC Supplier No. HYW017758S11MG
1,3-Propanediol-d8 is the deuterium-labeled analogue of 1,3-propanediol used as an isotope-labeled standard and reagent in analytical and synthetic research. It offers high isotopic enrichment and is provided in small, well-characterized quantities suitable for calibration, tracer studies, and method development.
  • Deuterium-labeled analogue for isotope tracing and standards.
  • High purity suitable for analytical applications (listed as 98.9%).
  • Suitable for GC-MS, NMR, and mass spectrometry workflows.
  • Provided in milligram-scale packages for standards and calibration.
  • Comes with documentation such as data sheet, certificate of analysis, and safety data sheet.