Learn More
Medchemexpress LLC 1,3-Propanedisulfonic acid, sodium salt (1:2) | 36589-58-9 | 98.0% | 100 MG
SDP
Catalog No. 5000413101
Encompass_Preferred
Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate. It is a negatively charged sulfonated inhibitor of fibrillogenesis that can be used in the treatment of amyloid A (AA) amyloidosis.
  • Orally available disodium salt form.
  • Negatively charged sulfonated inhibitor of fibrillogenesis.
  • Used in the treatment of amyloid A (AA) amyloidosis.
Catalog No. 50-004-13101 Supplier Medchemexpress LLC Supplier No. HYB1141100MG
Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate. It is a negatively charged sulfonated inhibitor of fibrillogenesis that can be used in the treatment of amyloid A (AA) amyloidosis.
  • Orally available disodium salt form.
  • Negatively charged sulfonated inhibitor of fibrillogenesis.
  • Used in the treatment of amyloid A (AA) amyloidosis.