Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Propanesultam | 5908-62-3 | MFCD09802220 | 121.16 | 250 MG
SDP

Catalog No. 5000449801
Encompass_Preferred

1,3-Propanesultam (1,1-Dioxoisothiazolidine) is a bioactive chemical suitable for the synthesis of active compounds. This product is intended for research use only and is not sold to patients. It has a solid appearance, ranging from colorless to off-white.

  • Purity ≥97.0%
  • Molecular formula C3H7NO2S
  • Molecular weight 121.16
  • For research use only

Catalog No. 50-004-49801 Supplier Medchemexpress LLC Supplier No. 250MGHYW001194
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Propanesultam (1,1-Dioxoisothiazolidine) is a bioactive chemical suitable for the synthesis of active compounds. This product is intended for research use only and is not sold to patients. It has a solid appearance, ranging from colorless to off-white.

  • Purity ≥97.0%
  • Molecular formula C3H7NO2S
  • Molecular weight 121.16
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.