Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-Pyridinylmethyl)piperazine hydrochloride

Catalog No. CDS002717 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002717 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002717 Supplier Sigma Aldrich Supplier No. CDS002717100MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 6-amino-1-methyluracil is an amino derivative of uracil and its standard molar enthalpy of combustion has been evaluated
  • Host-guest complexation of 6-amino-1-methyluracil with tetrapropoxycalix[4]arene has been reported.6-amino-1-methyluracil may be used in the preparation of 1,1α-di methyl-1h-spiro[pyrimido[4,5-b]quinoline-5,5α-pyrrolo[2,3-d]pyrimidine]-2,2α,4,4α,6α(1αh,3h,3αh,7αh,1αh)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid

Specifications

Molecular Formula C10H16ClN3
Linear Formula C10H16ClN3
MDL Number MFCD06801255
Quantity 25 g
Molecular Weight (g/mol) 213.71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.