Learn More
Medchemexpress LLC 1-(3-sulfopropyl)pyridinium hydroxide inner salt | 15471-17-7 | MFCD00064468 | 98.5% | 201.24 g·mol⁻¹ | C8H11NO3S | 25 G
SDP
Catalog No. 5000303563
Encompass_Preferred
NDSB-201 (1-(3-sulfopropyl)pyridinium hydroxide inner salt) is a zwitterionic sulfobetaine reagent used as a non-denaturing additive in biochemical and protein research. It is typically employed to improve solubility and stability of proteins during extraction, refolding, and assay workflows, while minimizing disruption to native structure.
  • Zwitterionic, non-denaturing additive for sensitive proteins.
  • Enhances protein solubility in aqueous buffers.
  • Facilitates protein refolding and recovery of activity.
  • Reduces aggregation during purification and assays.
  • High water solubility and compatibility with common buffers.
  • Provided at research-grade purity for reproducible results.
Catalog No. 50-003-03563 Supplier Medchemexpress LLC Supplier No. 25GHYW014837
NDSB-201 (1-(3-sulfopropyl)pyridinium hydroxide inner salt) is a zwitterionic sulfobetaine reagent used as a non-denaturing additive in biochemical and protein research. It is typically employed to improve solubility and stability of proteins during extraction, refolding, and assay workflows, while minimizing disruption to native structure.
  • Zwitterionic, non-denaturing additive for sensitive proteins.
  • Enhances protein solubility in aqueous buffers.
  • Facilitates protein refolding and recovery of activity.
  • Reduces aggregation during purification and assays.
  • High water solubility and compatibility with common buffers.
  • Provided at research-grade purity for reproducible results.