Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™

Catalog No. CC09509DA
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC09509DA 1 g Amber Glass Bottle
CC09509DE 5 g Amber Glass Bottle
CC09509EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC09509DA Supplier Thermo Scientific Chemicals Supplier No. CC09509DA
Add to Cart

Chemical Identifiers

CAS 14542-12-2
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD06200855
InChI Key JNHDLNXNYPLBMJ-UHFFFAOYSA-N
Synonym thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol
PubChem CID 2795213
SMILES OCC1=NC=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 1,3-thiazol-2-ylmethanol
Formula Weight 115.16
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-2-ylmethanol
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.