Learn More
Cayman Chemical 1-(3-Trifluoromethylphenyl)piperazine (hydrochloride), Purity: ≥98%, CAS: 76835-14-8, 10 mg
SDP
Catalog No. 502951530
Encompass_Preferred
An entactogenic drug which selectively promotes the release of serotonin; in combination with BZP, increases both serotonin and dopamine, mirroring the effects of MDMA; has been identified in party pills and powders and is intended for forensic applications
Catalog No. 50-295-1530 Supplier Cayman Chemical Supplier No. 1120510 MG
An entactogenic drug which selectively promotes the release of serotonin; in combination with BZP, increases both serotonin and dopamine, mirroring the effects of MDMA; has been identified in party pills and powders and is intended for forensic applications