Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-[3-(Trimethoxysilyl)propyl]urea

Catalog No. 440817100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-081-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-081-7100ML Supplier Sigma Aldrich Supplier No. 440817100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-[3-(Trimethoxysilyl)propyl]urea is a urea based surface modifying agent that is used for forming self-assembled monolayers (SAMs) on a variety of substrates. It is mainly utilized to immobilize the surface atoms by forming a ligand.1-[3-(Trimethoxysilyl)propyl]urea capped magnetic nanoparticles can be used as catalyst supports for immobilizing the surface enzymes and nanoparticles. It may be used in the silanization of ceria that finds potential applications as medical agents.

Specifications

CAS 23843-64-3
Density 1.15 g/mL (at 25°C)
Boiling Point 217°C to 250°C
Molecular Formula C7H18N2O4Si
Linear Formula (CH3O)3 Si(CH2)3 NHCONH2
Refractive Index n20/D 1.46 (literature)
MDL Number MFCD00216654
Quantity 100 mL
Synonym gamma-Ureidopropyltrimethoxysilane; 3-Ureidopropyltrimethoxysilane; N -[3-(Trimethoxysilyl)propyl]urea; Ureidopropyltrimethoxysilane
Molecular Weight (g/mol) 222.31
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.