Learn More
Optimize Technologies 1/32" OPTI-PEEK™ Sleeves for Fused Silica Tubing , 10pk
SDP
Catalog No. 502006192
Encompass_Preferred
OPTI-PEEK™ Sleeves allow 360um fused silica tubing to be used with EXP2 TI-LOK Fittings and Adapters.  Available in 2 and 10 packs. Dimensions: 1/32" OD x .015" ID x 1" LG.
Catalog No. 50-200-6192 Supplier Optimize Technologies Supplier No. 101305321
OPTI-PEEK™ Sleeves allow 360um fused silica tubing to be used with EXP2 TI-LOK Fittings and Adapters.  Available in 2 and 10 packs. Dimensions: 1/32" OD x .015" ID x 1" LG.