Learn More
Cayman Chemical 1 3BenzodIoxole5 6dIamIn 10mg
SDP
Catalog No. 502946190
Encompass_Preferred
A fluorescent derivatization reagent; has been commonly used for the derivatization of α-keto, neuraminic, and sialic acids for detection by fluorometric HPLC
Catalog No. 50-294-6190 Supplier Cayman Chemical Supplier No. 3430810 MG
A fluorescent derivatization reagent; has been commonly used for the derivatization of α-keto, neuraminic, and sialic acids for detection by fluorometric HPLC