Learn More
Cayman Chemical 1,3-Benzodioxole-5,6-diamine (hydrochloride), Purity: ≥98%, CAS: 81864-15-5, 50 mg
SDP
Catalog No. 502946847
Encompass_Preferred
A fluorescent derivatization reagent; has been commonly used for the derivatization of α-keto, neuraminic, and sialic acids for detection by fluorometric HPLC
Catalog No. 50-294-6847 Supplier Cayman Chemical Supplier No. 3430850 MG
A fluorescent derivatization reagent; has been commonly used for the derivatization of α-keto, neuraminic, and sialic acids for detection by fluorometric HPLC