Learn More
Cayman Chemical 1-(3-Chlorophenyl)piperazine (hydrochloride), Purity: ≥98%, CAS: 13078-15-4, 10 mg
SDP
Catalog No. 502954271
Encompass_Preferred
An analytical reference standard categorized as a piperazine; a metabolite of trazodone; has been found in 3,4-MDMA tablets seized by law enforcement; intended for research and forensic applications
Catalog No. 50-295-4271 Supplier Cayman Chemical Supplier No. 1394910 MG
An analytical reference standard categorized as a piperazine; a metabolite of trazodone; has been found in 3,4-MDMA tablets seized by law enforcement; intended for research and forensic applications