Learn More
Cayman Chemical 1 3DIdocosahanyl GlycRo 10mg
SDP
Catalog No. 502937903
Encompass_Preferred
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions
Catalog No. 50-293-7903 Supplier Cayman Chemical Supplier No. 2701110 MG
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions