Learn More
Cayman Chemical 1,3-Didocosahexaenoyl-2-Palmitoyl Glycerol, Purity: ≥98%, CAS: 197016-86-7, 10 mg
SDP
Catalog No. 502939415
Encompass_Preferred
A triacylglycerol; has been found in higher amounts than PDD in the Steller sea lion meat and harp seal oil and at lower amounts than PDD in the meat of a variety of fishes
Catalog No. 50-293-9415 Supplier Cayman Chemical Supplier No. 2698710 MG
A triacylglycerol; has been found in higher amounts than PDD in the Steller sea lion meat and harp seal oil and at lower amounts than PDD in the meat of a variety of fishes