Learn More
Cayman Chemical 1,3-Didocosanoyl-2-Oleoyl Glycerol, Purity: ≥95%, CAS: 77145-68-7, 10 mg
SDP
Catalog No. 502941193
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter; increases the solidification rate of and prevents fat bloom formation on dark chocolate when used as a seed material
Catalog No. 50-294-1193 Supplier Cayman Chemical Supplier No. 2727810 MG
A triacylglycerol; has been found in cocoa butter; increases the solidification rate of and prevents fat bloom formation on dark chocolate when used as a seed material