Learn More
Cayman Chemical 1 3DIdocosnoyl2Oleoyl Gly 50mg
SDP
Supplier:  Cayman Chemical 2727850 MG
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter; increases the solidification rate of and prevents fat bloom formation on dark chocolate when used as a seed material
Catalog No. 50-294-0732