Learn More
Cayman Chemical 1 3DIlInoelaIdoyl GlycRol 10mg
SDP
Catalog No. 502937996
Encompass_Preferred
A diacylglycerol; a collision-induced decomposition product of synthetic linoleic ammoniated triglycerides
Catalog No. 50-293-7996 Supplier Cayman Chemical Supplier No. 2698310 MG
A diacylglycerol; a collision-induced decomposition product of synthetic linoleic ammoniated triglycerides