Learn More
Cayman Chemical 1,3-Dilinoleoyl-2-Stearoyl Glycerol, Purity: ≥98%, CAS: 2190-18-3, 5 mg
SDP
Catalog No. 502941718
Encompass_Preferred
A triacylglycerol; has been found in soybean oil as well as transitional and mature human milk
Catalog No. 50-294-1718 Supplier Cayman Chemical Supplier No. 269715 MG
A triacylglycerol; has been found in soybean oil as well as transitional and mature human milk