Learn More
Cayman Chemical 1,3-Dilinoleoyl-2-Oleoyl Glycerol, Purity: ≥98%, CAS: 2190-22-9, 50 mg
SDP
Catalog No. 502939587
Encompass_Preferred
A triacylglycerol; has been found in grape seed, pumpkin seed, soybean, sunflower, and wheat germ oils
Catalog No. 50-293-9587 Supplier Cayman Chemical Supplier No. 2706450 MG
A triacylglycerol; has been found in grape seed, pumpkin seed, soybean, sunflower, and wheat germ oils