Learn More
Cayman Chemical 1 3DIlnoloyl2Stearoyl Gl 10mg
SDP
Catalog No. 502939408
Encompass_Preferred
A triacylglycerol; has been found in soybean oil as well as transitional and mature human milk
Catalog No. 50-293-9408 Supplier Cayman Chemical Supplier No. 2697110 MG
A triacylglycerol; has been found in soybean oil as well as transitional and mature human milk