Learn More
Cayman Chemical 1 3DImrIstol2EIcosapenta 10mg
SDP
Catalog No. 502939726
Encompass_Preferred
A triacylglycerol; the myristoyl groups in MEM do not affect the oxidative stability of EPA during long-term storage at 25°C
Catalog No. 50-293-9726 Supplier Cayman Chemical Supplier No. 2698610 MG
A triacylglycerol; the myristoyl groups in MEM do not affect the oxidative stability of EPA during long-term storage at 25°C