Learn More
Cayman Chemical 1,3-Dioctanoyl-2-Oleoyl Glycerol, Purity: ≥98%, CAS: 109796-58-9, 1 mg
SDP
Catalog No. 502934741
Encompass_Preferred
A triacylglycerol; reduces total cholesterol levels in both the plasma and aorta in a diet-induced hamster model of hypercholesterolemia
Catalog No. 50-293-4741 Supplier Cayman Chemical Supplier No. 268201 MG
A triacylglycerol; reduces total cholesterol levels in both the plasma and aorta in a diet-induced hamster model of hypercholesterolemia