Learn More
Cayman Chemical 1,3-Dioleoyl-2-Elaidoyl Glycerol, Purity: ≥95%, CAS: 110170-81-5, 1 mg
SDP
Catalog No. 502938306
Encompass_Preferred
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols
Catalog No. 50-293-8306 Supplier Cayman Chemical Supplier No. 292351 MG
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols