Learn More
Cayman Chemical 1,3-Dioleoyl-2-Elaidoyl Glycerol, Purity: ≥95%, CAS: 110170-81-5, 5 mg
SDP
Catalog No. 502937164
Encompass_Preferred
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols
Catalog No. 50-293-7164 Supplier Cayman Chemical Supplier No. 292355 MG
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols