Learn More
Cayman Chemical 1,3-Dioleoyl-2-Lauroyl Glycerol, Purity: ≥98%, CAS: 383189-71-7, 10 mg
SDP
Catalog No. 502941200
Encompass_Preferred
A triacylglycerol; has been found in human milk, butterfat, and palm kernel oil
Catalog No. 50-294-1200 Supplier Cayman Chemical Supplier No. 2728710 MG
A triacylglycerol; has been found in human milk, butterfat, and palm kernel oil