Learn More
Cayman Chemical 1,3-Dioleoyl-2-Linoleoyl Glycerol, Purity: ≥95%, CAS: 2190-19-4, 5 mg
SDP
Catalog No. 502941602
Encompass_Preferred
A triacylglycerol; has been found as a major molecular species in butterfat; has been found in ASO, as well as, to a lesser extent, in physical blends and interesterified products of ASO
Catalog No. 50-294-1602 Supplier Cayman Chemical Supplier No. 269725 MG
A triacylglycerol; has been found as a major molecular species in butterfat; has been found in ASO, as well as, to a lesser extent, in physical blends and interesterified products of ASO