Learn More
Cayman Chemical 1,3-Dioleoyl-2-Octanoyl Glycerol, Purity: ≥98%, CAS: 139979-77-4, 10 mg
SDP
Catalog No. 502932772
Encompass_Preferred
A triacylglycerol; been used to characterize the functionality of R. oryzae lipase for regiodistribution analysis of fats containing medium- and short-chain fatty acids
Catalog No. 50-293-2772 Supplier Cayman Chemical Supplier No. 2683610 MG
A triacylglycerol; been used to characterize the functionality of R. oryzae lipase for regiodistribution analysis of fats containing medium- and short-chain fatty acids