Learn More
Cayman Chemical 1,3-Dioleoyl-2-Palmitoyl Glycerol, Purity: ≥98%, CAS: 1716-07-0, 5 mg
SDP
Catalog No. 502935020
Encompass_Preferred
A triacylglycerol; one of the most common triacylglycerols found in human breast milk; levels are higher in colostrum than transitional and mature milk; levels of 1,3-dioleoyl-2-palmitoyl glycerol are higher in Western women compared with Chinese women
Catalog No. 50-293-5020 Supplier Cayman Chemical Supplier No. 268435 MG
A triacylglycerol; one of the most common triacylglycerols found in human breast milk; levels are higher in colostrum than transitional and mature milk; levels of 1,3-dioleoyl-2-palmitoyl glycerol are higher in Western women compared with Chinese women