Learn More
Cayman Chemical 1,3-Dioleoyl-2-Elaidoyl Glycerol, Purity: ≥95%, CAS: 110170-81-5, 25 mg
SDP
Catalog No. 502937192
Encompass_Preferred
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols
Catalog No. 50-293-7192 Supplier Cayman Chemical Supplier No. 2923525 MG
A triacylglycerol; has been used to study the effects of unsaturation and sn-2 substitution on the polymorphic behavior of triacylglycerols