Learn More
Cayman Chemical 1 3DIpalmItol2Starol Gl 10mg
SDP
Catalog No. 502938004
Encompass_Preferred
A triacylglycerol; levels decrease in rats with reduced food intake-induced energy depletion
Catalog No. 50-293-8004 Supplier Cayman Chemical Supplier No. 2698910 MG
A triacylglycerol; levels decrease in rats with reduced food intake-induced energy depletion