Learn More
Cayman Chemical 1 3DIpalmItoleoyl Glycerol 1mg
SDP
Catalog No. 502932948
Encompass_Preferred
A diacylglycerol with palmitoleic acid at the sn-1 and sn-3 positions; selectively inhibits α-glucosidase from S. cerevisiae over rat enzyme (IC50s = 4.45 and 9,326.5 μM, respectively); has been used in the formation of lipid bilayers to study the effects of membrane composition and valinomycin on membrane potential
Catalog No. 50-293-2948 Supplier Cayman Chemical Supplier No. 267971 MG
A diacylglycerol with palmitoleic acid at the sn-1 and sn-3 positions; selectively inhibits α-glucosidase from S. cerevisiae over rat enzyme (IC50s = 4.45 and 9,326.5 μM, respectively); has been used in the formation of lipid bilayers to study the effects of membrane composition and valinomycin on membrane potential