Learn More
Cayman Chemical 1,3-Dipalmitoleoyl Glycerol, Purity: ≥98%, CAS: 106352-09-4, 10 mg
SDP
Catalog No. 502932674
Encompass_Preferred
A diacylglycerol with palmitoleic acid at the sn-1 and sn-3 positions; selectively inhibits α-glucosidase from S. cerevisiae over rat enzyme (IC50s = 4.45 and 9,326.5 μM, respectively); has been used in the formation of lipid bilayers to study the effects of membrane composition and valinomycin on membrane potential
Catalog No. 50-293-2674 Supplier Cayman Chemical Supplier No. 2679710 MG
A diacylglycerol with palmitoleic acid at the sn-1 and sn-3 positions; selectively inhibits α-glucosidase from S. cerevisiae over rat enzyme (IC50s = 4.45 and 9,326.5 μM, respectively); has been used in the formation of lipid bilayers to study the effects of membrane composition and valinomycin on membrane potential