Learn More
Cayman Chemical 1,3-Dipalmitoyl-2-Oleoyl Glycerol, Purity: ≥95%, CAS: 2190-25-2, 10 mg
SDP
Catalog No. 502937763
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil
Catalog No. 50-293-7763 Supplier Cayman Chemical Supplier No. 2706610 MG
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil