Learn More
Cayman Chemical 1,3-Dipalmitoyl-2-Oleoyl Glycerol, Purity: ≥95%, CAS: 2190-25-2, 25 mg
SDP
Catalog No. 502938533
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil
Catalog No. 50-293-8533 Supplier Cayman Chemical Supplier No. 2706625 MG
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil