Learn More
Cayman Chemical 1 3DIpalmItoyl2oloyl Glyc 50mg
SDP
Catalog No. 502938316
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil
Catalog No. 50-293-8316 Supplier Cayman Chemical Supplier No. 2706650 MG
A triacylglycerol; has been found in cocoa butter, Chinese tallow butter, and the palm stearin fraction of palm oil