Learn More
Cayman Chemical 1,3-Dipalmitoyl-2-Stearoyl Glycerol, Purity: ≥98%, CAS: 2177-97-1, 100 mg
SDP
Catalog No. 502939750
Encompass_Preferred
A triacylglycerol; levels decrease in rats with reduced food intake-induced energy depletion
Catalog No. 50-293-9750 Supplier Cayman Chemical Supplier No. 26989100 MG
A triacylglycerol; levels decrease in rats with reduced food intake-induced energy depletion