Learn More
Cayman Chemical 1,3-Dipentadecanoyl Glycerol, Purity: ≥95%, CAS: 59891-25-7, 50 mg
SDP
Catalog No. 502941661
Encompass_Preferred
A diacylglycerol; has been used as an internal standard for the quantification of ceramides and diacylglycerols in rat soleus muscle by LC-MS/MS
Catalog No. 50-294-1661 Supplier Cayman Chemical Supplier No. 2699450 MG
A diacylglycerol; has been used as an internal standard for the quantification of ceramides and diacylglycerols in rat soleus muscle by LC-MS/MS