Learn More
Cayman Chemical 1 3DIstearoyl2lnoloyl Gl 10mg
SDP
Catalog No. 502939729
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter and in CBEs containing shea stearin and palm mid fraction
Catalog No. 50-293-9729 Supplier Cayman Chemical Supplier No. 2696710 MG
A triacylglycerol; has been found in cocoa butter and in CBEs containing shea stearin and palm mid fraction