Learn More
Cayman Chemical 1,3-Distearoyl-2-Oleoyl Glycerol, Purity: ≥98%, CAS: 2846-04-0, 5 mg
SDP
Catalog No. 502937412
Encompass_Preferred
A triacylglycerol; has been found in Kpangnana, Chinese tallow, and cocoa butters; increases the phagocytic function of reticulo-endothelial cells in mice at 5 and 10 mg per animal
Catalog No. 50-293-7412 Supplier Cayman Chemical Supplier No. 271635 MG
A triacylglycerol; has been found in Kpangnana, Chinese tallow, and cocoa butters; increases the phagocytic function of reticulo-endothelial cells in mice at 5 and 10 mg per animal