Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(3H-benzimidazole-5-carbonyl)-2,3,4,4a,9,9a-hexahydroindeno[2,1-b]pyridine-6-carbonitrile | 1303515-32-3 | 98.1% | 342.39 g/mol | C21H18N4O | 10MG
SDP

Catalog No. 5000236612
Encompass_Preferred

BI 187004 (VTP-34072) is a selective small-molecule inhibitor of 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) used in metabolic and diabetes research. It is supplied for laboratory research use only, has been evaluated in clinical pharmacology studies, and is characterized by a defined chemical composition and high supplier-reported purity.

  • Target: 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1).
  • Chemical formula C21H18N4O.
  • Molecular weight 342.39 g/mol.
  • Purity 98.1% (supplier listing).
  • For research use only; not for human or veterinary use.
  • Suitable for preclinical in vitro and in vivo research applications.

Catalog No. 50-002-36612 Supplier Medchemexpress LLC Supplier No. HY12380010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI 187004 (VTP-34072) is a selective small-molecule inhibitor of 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) used in metabolic and diabetes research. It is supplied for laboratory research use only, has been evaluated in clinical pharmacology studies, and is characterized by a defined chemical composition and high supplier-reported purity.

  • Target: 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1).
  • Chemical formula C21H18N4O.
  • Molecular weight 342.39 g/mol.
  • Purity 98.1% (supplier listing).
  • For research use only; not for human or veterinary use.
  • Suitable for preclinical in vitro and in vivo research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.