Learn More
NEST SCIENTIFIC INC 1.3 mL 96-Well Deep Well Plate, U-Bottom, Round Well, Non-Sterile,50/cs
Catalog No. NC3236104
Encompass_Preferred
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).
Catalog No. NC3236104 Supplier NEST SCIENTIFIC INC Supplier No. 507001
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).