Learn More
SAI Infusion Technologies 1.3ml Lithium Heparin Blood Tubes with Screw Cap
SDP
Catalog No. NC3589066
Encompass
1.3ml Lithium Heparin Blood Tube with Screw Cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.
Catalog No. NC3589066 Supplier SAI Infusion Technologies Supplier No. PMTSH1.3
1.3ml Lithium Heparin Blood Tube with Screw Cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.