Learn More
SAI Infusion Technologies 1.3ml Serum Blood Tubes with Screw Cap
SDP
Catalog No. NC3589052
Encompass
1.3ml Serum Blood Tube with Clotting Activator and Screw Cap. Ideal for collection of up to 1.3ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.
Catalog No. NC3589052 Supplier SAI Infusion Technologies Supplier No. PMTSS1.3
1.3ml Serum Blood Tube with Clotting Activator and Screw Cap. Ideal for collection of up to 1.3ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.