Learn More
Glass Expansion 1.3mm Ferrules
SDP
Supplier:  Glass Expansion 708030748
Encompass_Preferred
1.3mm Ferrules (PKT 10). Package of ten 1.3mm ferrules. Compatible with Glass Expansion's QA Kit (KT-1136), TruFlo Sample Monitors (501907281, 501907282, 501907283, 501907284, 501907285, 501907286), HydraMist Spray Chambers (KT-1160), Trident CT Internal Standard Mixing Kits (NC2046960), and Guardian In-Line Sample Filters (NC0897716). Suitable for use with Ratchet Connector 70-803-1350, and the following 1.3mm OD capillary PFA sample tubing STT-075 (0.75mm ID x 10m long), STT-050 (0.50mm ID x 10m long), STT-025 (0.25mm ID x 10m long) to make custom length sample lines terminated with a 1/4-28 Ratchet Connector. Swage Tool for Ratchet Connector, 50-100-1811, secures ferrule on capillary and allows desired capillary tube length to be terminated with appropriate ferrule and ratchet fitting. For 1/16” Ferrules see 70-803-0749.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3088