Learn More
Medchemexpress LLC 1-[(3R)-1-methyl-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]-3-(3-methylphenyl)urea | 00-00-0 | MFCD00869756 | 99.8% | 407.47 g/mol | C24H24N4O3 | 5 MG
SDP
Catalog No. 5000241111
Encompass_Preferred
L-365260 hemihydrate is an orally active, selective antagonist of non-peptide gastrin and brain cholecystokinin B (CCK-B) receptors. It binds competitively and stereoselectively, with reported Ki values of 1.9 nM (gastrin) and 2.0 nM (brain CCK), and is supplied for research use only.
  • Potent CCK-B receptor antagonist with Ki ~1.9-2.0 nM.
  • Orally active and stereoselective in receptor binding.
  • High chemical purity of 99.75%.
  • Solid, white to off-white appearance for easy handling.
  • Available in small research pack sizes: 5 mg, 10 mg, 50 mg.
  • Intended for research use only; not for human use.
Catalog No. 50-002-41111 Supplier Medchemexpress LLC Supplier No. HY106840A5MG
L-365260 hemihydrate is an orally active, selective antagonist of non-peptide gastrin and brain cholecystokinin B (CCK-B) receptors. It binds competitively and stereoselectively, with reported Ki values of 1.9 nM (gastrin) and 2.0 nM (brain CCK), and is supplied for research use only.
  • Potent CCK-B receptor antagonist with Ki ~1.9-2.0 nM.
  • Orally active and stereoselective in receptor binding.
  • High chemical purity of 99.75%.
  • Solid, white to off-white appearance for easy handling.
  • Available in small research pack sizes: 5 mg, 10 mg, 50 mg.
  • Intended for research use only; not for human use.